1-Brom-2,3-dichlorbenzen ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,3-Dichlorobromobenzene

B0980-25G B0980-5G
TCIAB0980-25GEA 288 DKK
TCIAB0980-25G TCIAB0980-5G
1-Brom-2,3-dichlorbenzen ≥99.0% (by GC)
1-Brom-2,3-dichlorbenzen
Formel: C₆H₃BrCl₂
Molekylvægt: 225.9 g/mol
Kogepunkt: 243°C
Smeltepunkt: 60
MDL Number: MFCD00000536
CAS-nummer: 56961-77-4
EINECS: 260-476-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fdaeb00185b9565fb6141bdaee57bea87046edd&vid=3cec91531832a1e8c80f49d2218608a45a2a8bc0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 59.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR