3-Brombenzoesyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: m-Bromobenzoic Acid
B0551-25G B0551-250G
TCIAB0551-25GEA 247 DKK
TCIAB0551-25G TCIAB0551-250G
3-Brombenzoesyre ≥98.0% (by GC, titration analysis)
3-Brombenzoesyre
Formel: C₇H₅BrO₂
Kogepunkt: 308 °C (1013 hPa)
Smeltepunkt: 155 °C
Massefylde: 1,845 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002487
CAS-nummer: 585-76-2
EINECS: 209-562-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55b2bad18b979eb96e8dd8b280ecfed7135cedf6&vid=7f88b7906c4abd123792f340a9bd6e05f9264cc6&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C
Solubility in Methanol almost transparency