3-Brombenzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B1227-100G
B1227-25G
B1227-5G
TCIAB1227-100GEA
1570
DKK
TCIAB1227-100G
TCIAB1227-25G
TCIAB1227-5G
3-Brombenzonitril ≥98.0% (by GC)
3-Brombenzonitril
Formel:
C₇H₄BrN Kogepunkt: 225°C Smeltepunkt: 37…40 °C Massefylde: 1,562 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001796 CAS-nummer: 6952-59-6 EINECS: 230-127-9 UN nummer: 3439 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 41.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...