Benz[a]anthracen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Benz(alpha)anthracen, 1,2-Benzanthracen
B0017-1G
B0017-25G
B0017-5G
TCIAB0017-1GEA
517
DKK
TCIAB0017-1G
TCIAB0017-25G
TCIAB0017-5G
Benz[a]anthracen ≥98.0% (by GC)
Benz[a]anthracen
Formel:
C₁₈H₁₂ Kogepunkt: 438°C Massefylde: 1,245 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003599 CAS-nummer: 56-55-3 EINECS: 200-280-6 UN nummer: 3077 ADR: 9,III Merck Index: 14,01062 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...