4-Methoxyanthranilsyre ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 2-Amino-4-methoxybenzoic acid
A2319-1G A2319-5G
TCIAA2319-1GEA 321 DKK
TCIAA2319-1G TCIAA2319-5G
4-Methoxyanthranilsyre ≥97.0% (by HPLC, titration analysis)
4-Methoxyanthranilsyre
Formel: C₈H₉NO₃
Molekylvægt: 167.16 g/mol
MDL Number: MFCD00667729
CAS-nummer: 4294-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d1f442b9c0733ae40ba14119245585d8de63b7b&vid=9d1f442b9c0733ae40ba14119245585d8de63b7b&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light red to Green powder to crystalline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %