3,5-Dichlorthioanisol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D3520-25G
D3520-5G
TCIAD3520-25GEA
796
DKK
TCIAD3520-25G
TCIAD3520-5G
3,5-Dichlorthioanisol ≥98.0% (by GC)
3,5-Dichlorthioanisol
Formel:
C₇H₆Cl₂S Kogepunkt: 145°C |
MDL Number:
MFCD00870149 CAS-nummer: 68121-46-0 |
|
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...