9,10-Diphenylanthracen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D1689-1G D1689-25G
TCIAD1689-1GEA 206 DKK
TCIAD1689-1G TCIAD1689-25G
9,10-Diphenylanthracen ≥98.0% (by GC)
9,10-Diphenylanthracene
Formel: C₂₆H₁₈
Molekylvægt: 330.43 g/mol
Kogepunkt: 485°C
Smeltepunkt: 245
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001253
CAS-nummer: 1499-10-1
EINECS: 216-105-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e7b29285e3ee156f2d79eaef70f16cbf10d2b5a&vid=1e02eaf50a0cdf0efcc8d38c3ccad85807aa08c1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR