9,10-Dimethylanthracen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D0252-1G D0252-10G
TCIAD0252-1GEA 394 DKK
TCIAD0252-1G TCIAD0252-10G
9,10-Dimethylanthracen ≥98.0% (by GC)
9,10-Dimethylanthracene
Formel: C₁₆H₁₄
Molekylvægt: 206.29 g/mol
Kogepunkt: 371
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001262
CAS-nummer: 781-43-1
EINECS: 212-308-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de5910b35eba092f682df61a8c24eef618f2561c&vid=1a2e4a3faa88ef7bb5c459a4197e84aa1e5078c0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 180.0 to 185.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR