2-tert-Butylanthracen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3541-1G B3541-5G
TCIAB3541-1GEA 559 DKK
TCIAB3541-1G TCIAB3541-5G
2-tert-Butylanthracen ≥98.0% (by GC)
2-tert-Butylanthracen
Formel: C₁₈H₁₈
Kogepunkt: 363 °C (1013 hPa)
Smeltepunkt: 149 °C
MDL Number: MFCD00003581
CAS-nummer: 18801-00-8
EINECS: 242-588-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ef3e02bf3260f556cafffc68530268739e8076b&vid=d5920b239eb80486789244e97032570665084852&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR