9,10-Dichloranthracen ≥96.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
D0327-1G D0327-10G
TCIAD0327-1GEA 132 DKK
TCIAD0327-1G TCIAD0327-10G
9,10-Dichloranthracen ≥96.0% (by GC)
9,10-Dichloroanthracene
Formel: C₁₄H₈Cl₂
Molekylvægt: 247.12 g/mol
Kogepunkt: 366
MDL Number: MFCD00001246
CAS-nummer: 605-48-1
EINECS: 210-087-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47e2e51ea26eeb9f1d8464c3a61ec13d9223372&vid=dddd433cb67c4c65a782e9da476a22cb4f37d7d5&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 96.0 %
Melting point 210.0 to 214.0 deg-C
Solubility in hot Toluene almost transparency