9,10-Dibromanthracen ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
D0166-25G D0166-500G D0166-100G
TCIAD0166-25GEA 370 DKK
TCIAD0166-25G TCIAD0166-500G TCIAD0166-100G
9,10-Dibromanthracen ≥98.0% (by GC)
9,10-Dibromoanthracene
Formel: C₁₄H₈Br₂
Molekylvægt: 336.03 g/mol
Kogepunkt: 427
MDL Number: MFCD00001244
CAS-nummer: 523-27-3
EINECS: 208-342-4
Merck Index: 14,03018
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19c7c2d3d10b03416bc356763976e7c83026acb9&vid=4f747a0e7ca043a470626201a14df611a4019d23&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Yellow to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 228.0 deg-C
Solubility in hot Toluene almost transparency