2-Methyl-1-hexen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

M0525-1G M0525-5G
TCIAM0525-1GEA 493 DKK
TCIAM0525-1G TCIAM0525-5G
2-Methyl-1-hexen ≥97.0% (by GC)
2-Methyl-1-hexene
Formel: C₇H₁₄
Kogepunkt: 92°C
Smeltepunkt: –103 °C
Massefylde: 0,697 g/cm³ (20 °C)
Flammepunkt: -6°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009486
CAS-nummer: 6094-02-6
EINECS: 228-041-1
UN nummer: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11e006b7334971efe93c64fffa5b33e01229986f&vid=0a1e30a0d3af6b5518fd40f4bf3ab345f2c56537&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR