3-Chlor-2-methylpropen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C0209-25ML C0209-500ML
TCIAC0209-25MLEA 140 DKK
TCIAC0209-25ML TCIAC0209-500ML
3-Chlor-2-methylpropen ≥98.0% (by GC)
3-Chloro-2-methylpropene
Formel: C₄H₇Cl
Molekylvægt: 90.55 g/mol
Kogepunkt: 73°C
Smeltepunkt: -80
Flammepunkt: –12°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000953
CAS-nummer: 563-47-3
EINECS: 209-251-2
UN nummer: 2554
ADR: 3 II
Merck Index: 14,02148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a060aa83c37f19a9d81410c6b149d8a321411872&vid=9a1b69422e23dd7495b8747ab81c7e12865d4a78&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR