cis-4-Octen ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
(Z)-4-Octene
O0135-5ML
O0135-1ML
TCIAO0135-5MLEA
1050
DKK
TCIAO0135-5ML
TCIAO0135-1ML
cis-4-Octen ≥95.0% (by GC)
cis-4-Octene
Formel:
C₈H₁₆ Molekylvægt: 112.22 g/mol Kogepunkt: 122°C Smeltepunkt: -118.7 Flammepunkt: 17°C |
MDL Number:
MFCD00067484 CAS-nummer: 7642-15-1 EINECS: 231-578-4 UN nummer: 3295 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...