Triallylamin ≥95.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T0332-25ML
T0332-500ML
TCIAT0332-25MLEA
132
DKK
TCIAT0332-25ML
TCIAT0332-500ML
Triallylamin ≥95.0% (by GC, titration analysis)
Triallylamin
Formel:
C₉H₁₅N Kogepunkt: 156°C Massefylde: 0,796 g/cm³ (20 °C) Flammepunkt: 39°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00026093 CAS-nummer: 102-70-5 EINECS: 203-048-2 UN nummer: 2610 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...