8-Brom-1-octen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3257-25G B3257-5G
TCIAB3257-25GEA 2510 DKK
TCIAB3257-25G TCIAB3257-5G
8-Brom-1-octen ≥98.0% (by GC)
8-Bromo-1-octene
Formel: C₈H₁₅Br
Molekylvægt: 191.11 g/mol
Kogepunkt: 92°C
Flammepunkt: 78°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000275
CAS-nummer: 2695-48-9
EINECS: 220-268-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01ca4cd07091f7c904cccdb19e4298ab7744afed&vid=1b414afba65a5cbe0beeed1bfb05c68d27a98729&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR