Squalan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H0096-25ML
H0096-500ML
H0096-100ML
TCIAH0096-25MLEA
140
DKK
TCIAH0096-25ML
TCIAH0096-500ML
TCIAH0096-100ML
Squalan ≥98.0% (by GC)
Squalan
Formel:
C₃₀H₆₂ Kogepunkt: 470 °C (1013 hPa) Smeltepunkt: –35 °C Massefylde: 0,758 g/cm³ (20 °C) Flammepunkt: 217°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008953 CAS-nummer: 111-01-3 EINECS: 203-825-6 Merck Index: 14,08767 |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...