Isobutylamin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
I0095-25ML
I0095-500ML
TCIAI0095-25MLEA
132
DKK
TCIAI0095-25ML
TCIAI0095-500ML
Isobutylamin ≥98.0% (by GC)
Isobutylamin
Formel:
C₄H₁₁N Kogepunkt: 66°C Smeltepunkt: –83°C Massefylde: 0,7344 g/cm³ (20 °C) Flammepunkt: –9°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008146 CAS-nummer: 78-81-9 EINECS: 201-145-4 UN nummer: 1214 ADR: 3,II Merck Index: 14,05132 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...