3-(Brommethyl)heptan ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2-ethylhexane, 2-Ethylhexylbromide, 3-(Bromomethyl)heptane

B0596-25ML B0596-500ML B0596-100ML
TCIAB0596-25MLEA 132 DKK
TCIAB0596-25ML TCIAB0596-500ML TCIAB0596-100ML
3-(Brommethyl)heptan ≥97.0% (by GC)
2-Ethylhexyl bromide
Formel: C₈H₁₇Br
Molekylvægt: 193.13 g/mol
Kogepunkt: 156°C
Flammepunkt: 69°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000220
CAS-nummer: 18908-66-2
EINECS: 242-659-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78cb7c7e43da79c97ff5dff88b54c24e29358a40&vid=11b765d859db9e5f9d76f41fa0614c2287f001fa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR