1-Brom-3-methylbutan ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Methylbutylbromide

I0504-25G I0504-500G
TCIAI0504-25GEA 353 DKK
TCIAI0504-25G TCIAI0504-500G
1-Brom-3-methylbutan ≥96.0% (by GC)
1-Bromo-3-methylbutane
Formel: C₅H₁₁Br
Molekylvægt: 151.05 g/mol
Kogepunkt: 121°C
Smeltepunkt: -112
Flammepunkt: 21°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000253
CAS-nummer: 107-82-4
EINECS: 203-522-9
UN nummer: 2341
ADR: 3 III
Merck Index: 14,05114
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc8f168b3bbc3a68053e11639b9b7fdc9be0d4f5&vid=fd58c834c7fb0e1e545428e130f2ede72df0a978&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR