Pentaerythrityl tetrabromid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1,3-Dibromo-2,2-bis(bromomethyl)propane, Tetrakis(bromomethyl)methane, Pentaerythrityltetrabromide

P0752-25G P0752-5G
TCIAP0752-25GEA 1190 DKK
TCIAP0752-25G TCIAP0752-5G
Pentaerythrityl tetrabromid ≥98.0% (by GC)
Pentaerythrityl tetrabromid
Formel: C₅H₈Br₄
Molekylvægt: 387.73 g/mol
Kogepunkt: 305
Smeltepunkt: 163
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000210
CAS-nummer: 3229-00-3
EINECS: 221-764-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e27072073d3094a04be3fc039ac188c81206ff15&vid=5da4cd5db7285a2f044327745f733af0695a5b0c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 159.0 to 163.0 deg-C
Solubility in Chloroform very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR