n-Undecan ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
U0002-25ML
U0002-500ML
TCIAU0002-25MLEA
181
DKK
TCIAU0002-25ML
TCIAU0002-500ML
n-Undecan ≥99.0% (by GC)
n-Undecane
Formel:
H₃C(CH₂)₉CH₃ Kogepunkt: 195°C Smeltepunkt: -26°C Massefylde: 0,7416 g/cm³ (20 °C) Flammepunkt: 65°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008959 CAS-nummer: 1120-21-4 EINECS: 214-300-6 UN nummer: 2330 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...