Rubrene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

T0561-1G T0561-100MG
TCIAT0561-1GEA 1020 DKK
TCIAT0561-1G TCIAT0561-100MG
Rubrene ≥98.0% (by GC)
Rubrene
Formel: C₄₂H₂₈
Smeltepunkt: 331°C
MDL Number: MFCD00003703
CAS-nummer: 517-51-1
EINECS: 208-242-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93e824ee9511d447cca2b0b432eccf8e11646ad9&vid=93e824ee9511d447cca2b0b432eccf8e11646ad9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR