1-Pentanthiol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
n-Pentanethiol, 1-Mercaptopentane, n-Pentyl mercaptan
P0053-25ML
P0053-500ML
TCIAP0053-25MLEA
296
DKK
TCIAP0053-25ML
TCIAP0053-500ML
1-Pentanthiol ≥98.0% (by GC, titration analysis)
1-Pentanthiol
Formel:
C₅H₁₂S Molekylvægt: 104.22 g/mol Kogepunkt: 126°C Smeltepunkt: -76 Flammepunkt: 18°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004907 CAS-nummer: 110-66-7 EINECS: 203-789-1 UN nummer: 1111 ADR: 3 II Merck Index: 14,00611 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Iodometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...