1,5-Pentandiol ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
P0050-25G
P0050-500G
TCIAP0050-25GEA
132
DKK
TCIAP0050-25G
TCIAP0050-500G
1,5-Pentandiol ≥97.0% (by GC)
1,5-Pentanediol
Formel:
C₅H₁₂O₂ Kogepunkt: 239°C Smeltepunkt: –16 °C Massefylde: 0,9941 g/cm³ (20 °C) Flammepunkt: 135°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002978 CAS-nummer: 111-29-5 EINECS: 203-854-4 Merck Index: 14,07117 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...