1-Octanol ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
n-Octanol
O0036-25ML
O0036-500ML
TCIAO0036-25MLEA
132
DKK
TCIAO0036-25ML
TCIAO0036-500ML
1-Octanol ≥99.0% (by GC)
1-Octanol
Formel:
H₃C(CH₂)₆CH₂OH Kogepunkt: 194°C Smeltepunkt: -16°C Massefylde: 0,8246 g/cm³ (20 °C) Flammepunkt: 83°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002988 CAS-nummer: 111-87-5 EINECS: 203-917-6 Merck Index: 14,06751 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...