Formaldehyd diethyl acetal ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Ethylal, Diethoxymethane
D0457-25ML
D0457-500ML
TCIAD0457-25MLEA
132
DKK
TCIAD0457-25ML
TCIAD0457-500ML
Formaldehyd diethyl acetal ≥99.0% (by GC)
Formaldehyd diethyl acetal
Formel:
C₅H₁₂O₂ Molekylvægt: 104.15 g/mol Kogepunkt: 88 Smeltepunkt: -66 Flammepunkt: -6 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009246 CAS-nummer: 462-95-3 EINECS: 207-330-6 UN nummer: 2373 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...