2-Nitropropan ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

N0249-25G N0249-500G N0249-100G
TCIAN0249-25GEA 386 DKK
TCIAN0249-25G TCIAN0249-500G TCIAN0249-100G
2-Nitropropan ≥95.0% (by GC)
2-Nitropropane
Formel: C₃H₇NO₂
Molekylvægt: 89.09 g/mol
Kogepunkt: 120°C
Smeltepunkt: –91°C
Flammepunkt: 24°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007397
CAS-nummer: 79-46-9
EINECS: 201-209-1
UN nummer: 2608
ADR: 3 III
Merck Index: 14,06628
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84bc0e9a271fa41efccc4fe402eb94f58a9cad6a&vid=0452c5354a59799552fc0fb6ae33bf943fdf6609&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR