Heptylamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-Heptylamine, 1-Aminoheptane
H0045-25ML
H0045-500ML
TCIAH0045-25MLEA
197
DKK
TCIAH0045-25ML
TCIAH0045-500ML
Heptylamin ≥98.0% (by GC, titration analysis)
Heptylamin
Formel:
H₃C(CH₂)₅CH₂NH₂ Kogepunkt: 157°C Smeltepunkt: –18 °C Massefylde: 0,7754 g/cm³ (20 °C) Flammepunkt: 35°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008244 CAS-nummer: 111-68-2 EINECS: 203-895-8 UN nummer: 2734 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...