Tripropylamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
T0522-25ML
T0522-500ML
TCIAT0522-25MLEA
132
DKK
TCIAT0522-25ML
TCIAT0522-500ML
Tripropylamin ≥98.0% (by GC, titration analysis)
Tripropylamin
Formel:
C₉H₂₁N Kogepunkt: 156°C Smeltepunkt: –94 °C Massefylde: 0,754 g/cm³ (20 °C) Flammepunkt: 41°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009363 CAS-nummer: 102-69-2 EINECS: 203-047-7 UN nummer: 2260 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...