1,2-Diiodethan ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D1025-25G D1025-100G
TCIAD1025-25GEA 345 DKK
TCIAD1025-25G TCIAD1025-100G
1,2-Diiodethan ≥97.0% (by GC)
1,2-Diiodoethane
Formel: C₂H₄I₂
Kogepunkt: 206 °C (1013 hPa)
Smeltepunkt: 80…82 °C
Massefylde: 2,132 g/cm³ (20 °C)
Opbevaringstemperatur: Fryser
MDL Number: MFCD00001093
CAS-nummer: 624-73-7
EINECS: 210-859-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e61419ac3af7a29821bda88b8fab098aef8f6a8d&vid=e61419ac3af7a29821bda88b8fab098aef8f6a8d&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR