1-Iodperfluordecan ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H0844-25G
H0844-5G
TCIAH0844-25GEA
616
DKK
TCIAH0844-25G
TCIAH0844-5G
1-Iodperfluordecan ≥98.0% (by GC)
1-Iodperfluordecan
Formel:
C₁₀F₂₁I Kogepunkt: 102°C |
MDL Number:
MFCD00001065 CAS-nummer: 423-62-1 EINECS: 207-030-5 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 65.0 to 68.0 deg-C |
Solubility in Hexane | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...