1,4-Diiodperfluorbutan ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Octafluoro-1,4-diiodobutane, 1,4-Diiodooctafluorobutane
Stabilization: Stabilised with copper chip
D2329-5G D2329-25G
TCIAD2329-5GEA 550 DKK
TCIAD2329-5G TCIAD2329-25G
1,4-Diiodperfluorbutan ≥98.0% (by GC) stabilised
1,4-Diiodperfluorbutan
Formel: C₄F₈I₂
Kogepunkt: 63°C
Smeltepunkt: –3 °C
Massefylde: 2,5466 g/cm³ (0 °C)
MDL Number: MFCD00042263
CAS-nummer: 375-50-8
EINECS: 206-788-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14cd29f9fc43f62d2649cae88f2f692c4c3800d1&vid=14cd29f9fc43f62d2649cae88f2f692c4c3800d1&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light yellow to Red clear liquid
Purity (GC) min. 98.0 %