1,3-Dichlorpropan
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D0399-25ML
D0399-250ML
TCIAD0399-25MLEA
279
DKK
TCIAD0399-25ML
TCIAD0399-250ML
1,3-Dichlorpropan
1,3-Dichloropropane
Formel:
ClH₂CCH₂CH₂Cl Kogepunkt: 122°C Massefylde: 1,1717 g/cm³ (20 °C) Flammepunkt: 21°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000999 CAS-nummer: 142-28-9 EINECS: 205-531-3 UN nummer: 1993 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...