Smørsyre ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Butansyre
B0754-25ML
B0754-500ML
TCIAB0754-25MLEA
132
DKK
TCIAB0754-25ML
TCIAB0754-500ML
Smørsyre ≥99.0% (by GC, titration analysis)
Smørsyre
Formel:
C₄H₈O₂ Kogepunkt: 163°C Smeltepunkt: –3 °C Massefylde: 0,958 g/cm³ (20 °C) Flammepunkt: 75°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00002814 CAS-nummer: 107-92-6 EINECS: 203-532-3 UN nummer: 2820 ADR: 8,III Merck Index: 14,01593 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...