Butylacetat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

A0024-25ML A0024-500ML
TCIAA0024-25MLEA 132 DKK
TCIAA0024-25ML TCIAA0024-500ML
Butylacetat ≥99.0% (by GC)
Butylacetat
Formel: CH₃COO(CH₂)₃CH₃
Molekylvægt: 116.16 g/mol
Kogepunkt: 126°C
Smeltepunkt: -76
Flammepunkt: 28°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009445
CAS-nummer: 123-86-4
EINECS: 204-658-1
UN nummer: 1123
ADR: 3 III
Merck Index: 14,01535
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8b3adbd5bc57ba571aafcf4e56d9ae4e5c2fbef&vid=1fb9dc157824122d110c3559fcaca4b12d9386ee&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR