Isobutylacetat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methylpropyl acetate

A0034-25ML A0034-500ML
TCIAA0034-25MLEA 132 DKK
TCIAA0034-25ML TCIAA0034-500ML
Isobutylacetat ≥99.0% (by GC)
Isobutylacetat
Formel: C₆H₁₂O₂
Molekylvægt: 116.16 g/mol
Kogepunkt: 117°C
Smeltepunkt: -99
Flammepunkt: 21°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008932
CAS-nummer: 110-19-0
EINECS: 203-745-1
UN nummer: 1213
ADR: 3 II
Merck Index: 14,05130
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7e208f20136ed222d6d65b05ab5dd76117eaa2e&vid=ed2986b5d5c9ac4a424f65f0baf70fe63dc2d586&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR