2,4-Pentandion ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: ACAC, 2,4-Pentanedione

P0052-25ML P0052-500ML
TCIAP0052-25MLEA 132 DKK
TCIAP0052-25ML TCIAP0052-500ML
2,4-Pentandion ≥99.0% (by GC)
Acetylacetone
Formel: CH₃COCH₂COCH₃
Kogepunkt: 138°C
Smeltepunkt: –23°C
Massefylde: 0,96945 g/cm³ (20 °C)
Flammepunkt: 34°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008787
CAS-nummer: 123-54-6
EINECS: 204-634-0
UN nummer: 2310
ADR: 3,III
Merck Index: 14,00081
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78079845ff80daac31c096a1fed9ad46c0e5ee31&vid=c66f098bd40083889ce233eb2fa54f111948e6ce&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR