2,3-Pentandion ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Pentane-2,3-dione

P0051-25ML P0051-500ML P0051-100ML
TCIAP0051-25MLEA 222 DKK
TCIAP0051-25ML TCIAP0051-500ML TCIAP0051-100ML
2,3-Pentandion ≥97.0% (by GC)
2,3-Pentanedione
Formel: C₅H₈O₂
Kogepunkt: 108°C
Smeltepunkt: –52 °C
Massefylde: 0,96 g/cm³ (20 °C)
Flammepunkt: 19°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00009313
CAS-nummer: 600-14-6
EINECS: 209-984-8
UN nummer: 1224
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0496ba75a12129ede1716609ad70fbae180fbc8f&vid=c98bb4e5f203001ac6850b6a79f77142ad34afc8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR