2-Pentylfuran ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Amylfuran
P1209-25ML
P1209-5ML
TCIAP1209-25MLEA
1230
DKK
TCIAP1209-25ML
TCIAP1209-5ML
2-Pentylfuran ≥98.0% (by GC)
2-Pentylfuran
Formel:
C₉H₁₄O Kogepunkt: 155°C Massefylde: 0,838 g/cm³ (19 °C) Flammepunkt: 46°C |
MDL Number:
MFCD00036497 CAS-nummer: 3777-69-3 EINECS: 223-234-7 UN nummer: 1993 ADR: 3,III |
|
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Appearance | Colorless to Light yellow to Light orange clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...