(-)-1,4-dithio-L-threitol ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: L-Cleland's Reagent, L-threo-1,4-Dimercaptobutane-2,3-diol, (2R,3R)-1,4-Dimercaptobutane-2,3-diol, L-Dithiothreitol, L-DTT, [R-(R*,R*)]-1,4-Dimercapto-2,3-butandiol, (2R,3R)-(-)-1,4-Dimercapto-2,3-butanediol, (2R,3R)-(-)-2,3-Dihydroxy-1,4-butanedithiol

D1589-1G D1589-5G
TCIAD1589-1GEA 1560 DKK
TCIAD1589-1G TCIAD1589-5G
(-)-1,4-dithio-L-threitol ≥95.0% (by titrimetric analysis)
(-)-1,4-dithio-L-threitol
Formel: C₄H₁₀O₂S₂
Molekylvægt: 154.25 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00064305
CAS-nummer: 16096-97-2
EINECS: 240-263-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0928a6c002e9d4059a004c47187253096555f125&vid=0928a6c002e9d4059a004c47187253096555f125&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 48.0 to 53.0 deg-C
Specific rotation [a]20/D +22.0 to +28.0 deg (C=2.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR