3-Hydroxy-2-nitro-6-picolin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Methyl-2-nitropyridin-3-ol, 6-Methyl-2-nitro-3-pyridone, 3-Hydroxy-6-methyl-2-nitropyridine, 6-Metil-2-nitropyridine-3-ol
H0762-25G
H0762-5G
TCIAH0762-25GEA
1450
DKK
TCIAH0762-25G
TCIAH0762-5G
3-Hydroxy-2-nitro-6-picolin ≥98.0% (by GC)
3-Hydroxy-2-nitro-6-picolin
Formel:
C₆H₆N₂O₃ Molekylvægt: 154.13 g/mol |
MDL Number:
MFCD00006259 CAS-nummer: 15128-90-2 EINECS: 239-192-8 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 109.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...