2-Hydroxy-5-nitro-6-picolin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Hydroxy-3-nitro-2-picoline, 6-Hydroxy-2-methyl-3-nitropyridine, 6-Methyl-5-nitro-2-pyridone, 2-Hydroxy-6-methyl-5-nitropyridine, 2-Methyl-3-nitro-6-pyridone
H1160-1G
H1160-5G
TCIAH1160-1GEA
263
DKK
TCIAH1160-1G
TCIAH1160-5G
2-Hydroxy-5-nitro-6-picolin ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-nitro-6-picolin
Formel:
C₆H₆N₂O₃ |
MDL Number:
MFCD00092010 CAS-nummer: 28489-45-4 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 233.0 to 238.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...