2-Hydroxy-5-nitro-4-picolin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Hydroxy-4-methyl-5-nitropyridine, 4-Methyl-5-nitro-2-pyridone

H1050-1G H1050-5G
TCIAH1050-1GEA 189 DKK
TCIAH1050-1G TCIAH1050-5G
2-Hydroxy-5-nitro-4-picolin ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-nitro-4-picolin
Formel: C₆H₆N₂O₃
Molekylvægt: 154.13 g/mol
MDL Number: MFCD00010690
CAS-nummer: 21901-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7deb3e0909a15da7ccf1492e907e25b056491ae7&vid=7deb3e0909a15da7ccf1492e907e25b056491ae7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR