1,3-Diaminopropan, 2 HCl ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Propane-1,3-diamine dihydrochloride, 1,3-Propanediamine dihydrochloride, Trimethylenediamine dihydrochloride
T0613-25G T0613-100G
TCIAT0613-25GEA 370 DKK
TCIAT0613-25G TCIAT0613-100G
1,3-Diaminopropan, 2 HCl ≥97.0% (by titrimetric analysis)
1,3-Diaminopropan, 2 HCl
Formel: C₃H₁₂Cl₂N₂
Molekylvægt: 147.05 g/mol
MDL Number: MFCD00012525
CAS-nummer: 10517-44-9
EINECS: 234-051-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d44ff25251f62c94b6e1698cc72e2cb27e748976&vid=d44ff25251f62c94b6e1698cc72e2cb27e748976&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Solubility in Water very faint turbidity