2-Amino-5-nitro-4-picolin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Methyl-5-nitro-2-pyridylamine, 4-Methyl-5-nitropyridin-2-amine, 2-Amino-4-methyl-5-nitropyridine
A1638-1G
A1638-5G
TCIAA1638-1GEA
181
DKK
TCIAA1638-1G
TCIAA1638-5G
2-Amino-5-nitro-4-picolin ≥98.0% (by GC, titration analysis)
2-Amino-5-nitro-4-picolin
Formel:
C₆H₇N₃O₂ Smeltepunkt: 220 °C |
MDL Number:
MFCD00010692 CAS-nummer: 21901-40-6 |
|
Specifikationer Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 222.0 to 226.0 deg-C |
Solubility in hot Pyridine | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...