2-Amino-5-nitro-3-picolin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-3-methyl-5-nitropyridine, 3-Methyl-5-nitro-2-pyridylamine

A1891-1G A1891-5G
TCIAA1891-1GEA 140 DKK
TCIAA1891-1G TCIAA1891-5G
2-Amino-5-nitro-3-picolin ≥98.0% (by GC, titration analysis)
2-Amino-5-nitro-3-picolin
Formel: C₆H₇N₃O₂
Molekylvægt: 153.14 g/mol
Smeltepunkt: 255°C
MDL Number: MFCD03095060
CAS-nummer: 18344-51-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6544f3d1ef5891973ba2acdcf15a070714a0cfe6&vid=6544f3d1ef5891973ba2acdcf15a070714a0cfe6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR