2,4,6-Tris(dimethylamino-methyl)-phenol ≥80.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T0526-25G T0526-500G
TCIAT0526-25GEA 132 DKK
TCIAT0526-25G TCIAT0526-500G
2,4,6-Tris(dimethylamino-methyl)-phenol ≥80.0% (by GC)
2,4,6-Tris(dimethylamino-methyl)-phenol
Formel: C₁₅H₂₇N₃O
Kogepunkt: 135°C
Massefylde: 0,974 g/cm³ (15 °C)
MDL Number: MFCD00008330
CAS-nummer: 90-72-2
EINECS: 202-013-9
UN nummer: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cae4a8195c3e104bba3e90b46db0aa72fb8c25&vid=13cae4a8195c3e104bba3e90b46db0aa72fb8c25&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 80.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR