2-Fluorbenzensulfonamid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0557-25G F0557-5G
TCIAF0557-25GEA 1760 DKK
TCIAF0557-25G TCIAF0557-5G
2-Fluorbenzensulfonamid ≥98.0% (by GC)
2-Fluorbenzensulfonamid
Formel: C₆H₆FNO₂S
MDL Number: MFCD00042283
CAS-nummer: 30058-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ea990e5c240b3ecd8d902cedfceb505b0619c0&vid=32ea990e5c240b3ecd8d902cedfceb505b0619c0&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 158.0 to 163.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR