2-Fluor-5-trifluormethylphenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-Fluor-2-hydroxy-4-(trifluormethyl)benzol, 2-Fluoro-5-trifluoromethylphenol
F0780-25G
F0780-5G
TCIAF0780-25GEA
1620
DKK
TCIAF0780-25G
TCIAF0780-5G
2-Fluor-5-trifluormethylphenol ≥98.0% (by GC, titration analysis)
2-Fluor-5-trifluormethylphenol
Formel:
C₇H₄F₄O Molekylvægt: 180.1 g/mol Kogepunkt: 146 Massefylde: 1.436 Flammepunkt: 57°C |
MDL Number:
MFCD00061286 CAS-nummer: 141483-15-0 UN nummer: 1993 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...